Product Details

SNP ID
rs139343089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92222614 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATACAGAAGACCTTGGTGGGTGAG[C/T]CCATTCTGCCCAAGTGGGTTTTCAA
Phenotype
MIM: 600709
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IARS PubMed Links

Gene Details

Gene
IARS
Gene Name
isoleucyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002161.5 3935 Silent Mutation GGA,GGG G1204G NP_002152.2
NM_013417.3 3935 Silent Mutation GGA,GGG G1204G NP_038203.2

View Full Product Details