Product Details

SNP ID
rs139459217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137192669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACTCAAATGTGGTCTGCAGGCTT[C/T]TGTCGTTGAAGGAGATCTTCATCTG
Phenotype
MIM: 606946 MIM: 610882 MIM: 613354
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC2 PubMed Links

Gene Details

Gene
ANAPC2
Gene Name
anaphase promoting complex subunit 2
There are no transcripts associated with this gene.

Gene
SSNA1
Gene Name
SS nuclear autoantigen 1
There are no transcripts associated with this gene.

Gene
TPRN
Gene Name
taperin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128228.2 1748 Missense Mutation AAA,AGA K583R NP_001121700.2

View Full Product Details