Product Details
- SNP ID
-
rs139749463
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21385095 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGATCTGCTGGATCATCTCATGGA[A/G]GACAGGGATGGTTTCAGCCTTTTGG
- Phenotype
-
MIM: 147562
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA2
PubMed Links
Gene Details
- Gene
- IFNA2
- Gene Name
- interferon alpha 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000605.3 |
303 |
Missense Mutation |
CTC,TTC |
L79F |
NP_000596.2 |
View Full Product Details