Product Details
- SNP ID
-
rs139758508
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128260505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCTGCAGCTCAGCCAGCTGCTCC[C/T]TGAGCTCCCGGTTCTGGGAGAGTGC
- Phenotype
-
MIM: 602377
MIM: 602580
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNM1
PubMed Links
Gene Details
- Gene
- DNM1
- Gene Name
- dynamin 1
There are no transcripts associated with this gene.
- Gene
- GOLGA2
- Gene Name
- golgin A2
View Full Product Details