Product Details

SNP ID
rs139868448
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34088431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAAAGAGTTTCGTTCCAGACGA[A/G]TCGTAGCAGTGGGTGTAAACAGCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF12 PubMed Links

Gene Details

Gene
DCAF12
Gene Name
DDB1 and CUL4 associated factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015397.3 1450 Silent Mutation GAC,GAT D427D NP_056212.1
XM_011517843.1 1450 Silent Mutation GAC,GAT D231D XP_011516145.1

View Full Product Details