Product Details

SNP ID
rs140534069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120757686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAGGAGCTCTGGGTAATTTTCT[C/G]CATAAGTTTTTATTACTGTTAGATT
Phenotype
MIM: 609071
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXW2 PubMed Links

Gene Details

Gene
FBXW2
Gene Name
F-box and WD repeat domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012164.3 8476 UTR 3 NP_036296.2
XM_005251910.3 8476 Intron XP_005251967.1
XM_005251913.2 8476 Intron XP_005251970.1
XM_006717049.3 8476 Intron XP_006717112.1
XM_006717051.2 8476 Intron XP_006717114.1
XM_011518518.1 8476 Intron XP_011516820.1
XM_017014615.1 8476 Intron XP_016870104.1
XM_017014616.1 8476 Intron XP_016870105.1
XM_017014617.1 8476 Intron XP_016870106.1

View Full Product Details