Product Details

SNP ID
rs141636965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133537419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCACCCAGGACAACAGCCCAAC[A/G]TCCAATAGCCTGGAGGGGGGCACCG
Phenotype
MIM: 612277
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTSL2 PubMed Links

Gene Details

Gene
ADAMTSL2
Gene Name
ADAMTS like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145320.1 320 Silent Mutation ACA,ACG T35T NP_001138792.1
NM_014694.3 320 Silent Mutation ACA,ACG T35T NP_055509.2
XM_005272237.3 320 Silent Mutation ACA,ACG T144T XP_005272294.1
XM_005272238.3 320 Intron XP_005272295.1
XM_005272239.2 320 Silent Mutation ACA,ACG T35T XP_005272296.1
XM_011519241.2 320 Intron XP_011517543.2
XM_011519242.2 320 Intron XP_011517544.1

View Full Product Details