Product Details

SNP ID
rs141858334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69046398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGCTTTCAGAGTTCGAACCAAC[A/G]TGGCCTCAACCAGATTTGGAATGTC
Phenotype
MIM: 606829
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FXN PubMed Links

Gene Details

Gene
FXN
Gene Name
frataxin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000144.4 399 Missense Mutation CAT,CGT H60R NP_000135.2
NM_001161706.1 399 Missense Mutation CAT,CGT H60R NP_001155178.1
NM_181425.2 399 Missense Mutation CAT,CGT H60R NP_852090.1

View Full Product Details