Product Details

SNP ID
rs142013070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104617494 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATGGGCTGAGCAGGTAGAGAA[A/G]GCTTTGCTTCTCCCCTCAGAGGAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR13C9 PubMed Links

Gene Details

Gene
OR13C9
Gene Name
olfactory receptor family 13 subfamily C member 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001956.1 711 Silent Mutation GCC,GCT A237A NP_001001956.1

View Full Product Details