Product Details

SNP ID
rs142176550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135933258 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAAAGTTTATAAGCAATAAGATC[A/T]GAGAGCAGGAGCAGCTGCAGCACCT
Phenotype
MIM: 608129
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UBAC1 PubMed Links

Gene Details

Gene
UBAC1
Gene Name
UBA domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016172.2 1578 UTR 3 NP_057256.2

View Full Product Details