Product Details
- SNP ID
-
hCV174779602
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:68419816 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTGCAGTGGATGGCCTTTTCCC[A/C]TCTTGACAGAATTCTGTTAGAGCCT
- Phenotype
-
MIM: 600981
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PGM5
PubMed Links
Gene Details
- Gene
- PGM5
- Gene Name
- phosphoglucomutase 5
There are no transcripts associated with this gene.
View Full Product Details