Product Details

SNP ID
rs142411256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137244182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACTCTACTCACCGGGGATGAAC[C/G]CCATGTAGAAGGGGATCAGGCCTTG
Phenotype
MIM: 609826 MIM: 602660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001710.2 1771 Missense Mutation GCG,GGG A285G NP_001001710.1
XM_006717108.2 1771 Missense Mutation GCG,GGG A269G XP_006717171.2
XM_011518694.1 1771 Missense Mutation GCG,GGG A320G XP_011516996.1
XM_011518695.1 1771 Missense Mutation GCG,GGG A282G XP_011516997.1
XM_011518696.2 1771 Intron XP_011516998.1
XM_011518697.1 1771 Intron XP_011516999.1
XM_017014718.1 1771 Missense Mutation GCG,GGG A198G XP_016870207.1
Gene
SLC34A3
Gene Name
solute carrier family 34 member 3
There are no transcripts associated with this gene.

Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
There are no transcripts associated with this gene.

Gene
TUBB4B
Gene Name
tubulin beta 4B class IVb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006088.5 1771 Intron NP_006079.1

View Full Product Details