Product Details

SNP ID
rs142544785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34616105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGTGCAACCTGGGAAAGGATAA[A/C]CTGCTCCTCTAAAGCAAAAATGGAC
Phenotype
MIM: 607387
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARID3C PubMed Links

Gene Details

Gene
ARID3C
Gene Name
AT-rich interaction domain 3C
There are no transcripts associated with this gene.

Gene
DCTN3
Gene Name
dynactin subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281425.1 336 Intron NP_001268354.1
NM_001281426.1 336 Missense Mutation GTT,TTT V93F NP_001268355.1
NM_001281427.1 336 Missense Mutation GTT,TTT V93F NP_001268356.1
NM_007234.4 336 Missense Mutation GTT,TTT V93F NP_009165.1
NM_024348.3 336 Missense Mutation GTT,TTT V93F NP_077324.1
Gene
RPP25L
Gene Name
ribonuclease P/MRP subunit p25 like
There are no transcripts associated with this gene.

View Full Product Details