Product Details

SNP ID
rs143366138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33442093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCCAGCTTCACATTCTCTTCCT[C/T]GTTGGAGGGTGGGGGCTGCTCCAGG
Phenotype
MIM: 600170
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP3 PubMed Links

Gene Details

Gene
AQP3
Gene Name
aquaporin 3 (Gill blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318144.1 932 Missense Mutation CAA,CGA Q204R NP_001305073.1
NM_004925.4 932 Missense Mutation AAG,GAG K277E NP_004916.1

View Full Product Details