Product Details

SNP ID
rs143416740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136412154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAACCACATTGGATAATGGGCTT[C/T]GCGTGGCATCTCAGAATAAGTTTGG
Phenotype
MIM: 613036
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PMPCA PubMed Links

Gene Details

Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282944.1 240 Intron NP_001269873.1
NM_001282946.1 240 Intron NP_001269875.1
NM_015160.2 240 Intron NP_055975.1
XM_005266059.4 240 Missense Mutation CGC,TGC R77C XP_005266116.1
XM_011518417.2 240 Intron XP_011516719.1
XM_017014543.1 240 Intron XP_016870032.1
Gene
SDCCAG3
Gene Name
serologically defined colon cancer antigen 3
There are no transcripts associated with this gene.

View Full Product Details