Product Details

SNP ID
rs143554180
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136941288 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGCTGACGTCCAGGAAGATGA[A/T]GAAGCACTCGTCGTTGGGCGTGTAG
Phenotype
MIM: 120930 MIM: 609072 MIM: 612905
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C8G PubMed Links

Gene Details

Gene
C8G
Gene Name
complement component 8, gamma polypeptide
There are no transcripts associated with this gene.

Gene
FBXW5
Gene Name
F-box and WD repeat domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018998.3 1600 Missense Mutation ATC,TTC I474F NP_061871.1
XM_005266089.2 1600 Missense Mutation ATC,TTC I484F XP_005266146.2
XM_011518789.1 1600 Missense Mutation ATC,TTC I466F XP_011517091.1
XM_017014812.1 1600 Missense Mutation ATC,TTC I456F XP_016870301.1
Gene
LCN12
Gene Name
lipocalin 12
There are no transcripts associated with this gene.

View Full Product Details