Product Details

SNP ID
rs143791301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:26994971 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAACATTGTGCTTGTGCTTTAA[A/C]TTGCTATGAAATTTTAAGGAAATTT
Phenotype
MIM: 608040
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IFT74 PubMed Links

Gene Details

Gene
IFT74
Gene Name
intraflagellar transport 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099222.1 1774 Intron NP_001092692.1
NM_001099223.1 1774 Intron NP_001092693.1
NM_001099224.1 1774 Intron NP_001092694.1
NM_025103.2 1774 Intron NP_079379.2
XM_011518035.2 1774 Intron XP_011516337.1
XM_011518036.2 1774 Intron XP_011516338.1
XM_017015163.1 1774 Intron XP_016870652.1
XM_017015164.1 1774 Intron XP_016870653.1
Gene
LRRC19
Gene Name
leucine rich repeat containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022901.2 1774 UTR 3 NP_075052.1

View Full Product Details