Product Details

SNP ID
rs143961573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137050514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACTTGGGTGGAAAAGCCCCTCG[A/G]CCAGCAGGGGTGGAAGCCGTGGGTC
Phenotype
MIM: 602012 MIM: 605798
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENTPD2 PubMed Links

Gene Details

Gene
ENTPD2
Gene Name
ectonucleoside triphosphate diphosphohydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001246.3 853 Missense Mutation CCG,TCG P267S NP_001237.1
NM_203468.2 853 Missense Mutation CCG,TCG P267S NP_982293.1
XM_011519212.2 853 Missense Mutation CCG,TCG P164S XP_011517514.1
Gene
NPDC1
Gene Name
neural proliferation, differentiation and control 1
There are no transcripts associated with this gene.

View Full Product Details