Product Details

SNP ID
rs144337990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:81991342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGCAGGCCATGAATCCCATTGATT[C/G]TTGTGCTCGTCATCACGGACCACCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPATA31D1 PubMed Links

Gene Details

Gene
SPATA31D1
Gene Name
SPATA31 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001670.2 897 Missense Mutation TCT,TGT S291C NP_001001670.1
XM_017014710.1 897 Missense Mutation TCT,TGT S254C XP_016870199.1

View Full Product Details