Product Details

SNP ID
rs144776498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125149695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAAAGGAAATATGGCGTTGTC[A/G]TTCTGGGAGGAATAACACGTTCTGA
Phenotype
MIM: 612725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP6C PubMed Links

Gene Details

Gene
PPP6C
Gene Name
protein phosphatase 6 catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123355.1 914 Missense Mutation ACG,ATG T336M NP_001116827.1
NM_001123369.1 914 Missense Mutation ACG,ATG T277M NP_001116841.1
NM_002721.4 914 Missense Mutation ACG,ATG T299M NP_002712.1
XM_011518847.2 914 Missense Mutation ACG,ATG T287M XP_011517149.1
Gene
SCAI
Gene Name
suppressor of cancer cell invasion
There are no transcripts associated with this gene.

View Full Product Details