Product Details

SNP ID
rs146518581
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137192674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATGTGGTCTGCAGGCTTTTGTC[A/G]TTGAAGGAGATCTTCATCTGGGAGT
Phenotype
MIM: 606946 MIM: 610882 MIM: 613354
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANAPC2 PubMed Links

Gene Details

Gene
ANAPC2
Gene Name
anaphase promoting complex subunit 2
There are no transcripts associated with this gene.

Gene
SSNA1
Gene Name
SS nuclear autoantigen 1
There are no transcripts associated with this gene.

Gene
TPRN
Gene Name
taperin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128228.2 1743 Silent Mutation AAC,AAT N581N NP_001121700.2

View Full Product Details