Product Details

SNP ID
rs146787265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125148237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAGTGGGTTTTCAAAACTACCT[A/G]AACAGATGCTAAAGTGCTTTATTAA
Phenotype
MIM: 612725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP6C PubMed Links

Gene Details

Gene
PPP6C
Gene Name
protein phosphatase 6 catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123355.1 2372 UTR 3 NP_001116827.1
NM_001123369.1 2372 UTR 3 NP_001116841.1
NM_002721.4 2372 UTR 3 NP_002712.1
XM_011518847.2 2372 UTR 3 XP_011517149.1
Gene
SCAI
Gene Name
suppressor of cancer cell invasion
There are no transcripts associated with this gene.

View Full Product Details