Product Details

SNP ID
rs147954119
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:105662661 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAGGTATATCAACTTCTTGGT[C/T]AAGGTCTTGGGGGAGCAAAGCCTGC
Phenotype
MIM: 607440 MIM: 186855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FKTN PubMed Links

Gene Details

Gene
FKTN
Gene Name
fukutin
There are no transcripts associated with this gene.

Gene
TAL2
Gene Name
TAL bHLH transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005421.2 205 Silent Mutation GTC,GTT V55V NP_005412.1

View Full Product Details