Product Details

SNP ID
rs149055087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136410688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAAGATGGCGGCTGTGGTGCTGG[A/C]GGCGACGCGGTTGCTGCGGGGCTCG
Phenotype
MIM: 613036
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PMPCA PubMed Links

Gene Details

Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282944.1 31 Intron NP_001269873.1
NM_001282946.1 31 Intron NP_001269875.1
NM_015160.2 31 Intron NP_055975.1
XM_005266059.4 31 Missense Mutation GAG,GCG E7A XP_005266116.1
XM_011518417.2 31 Intron XP_011516719.1
XM_017014543.1 31 Intron XP_016870032.1
Gene
SDCCAG3
Gene Name
serologically defined colon cancer antigen 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039707.1 31 Intron NP_001034796.1
NM_001039708.1 31 Intron NP_001034797.1
NM_006643.3 31 Intron NP_006634.3
XM_005266050.3 31 Intron XP_005266107.1
XM_005266051.3 31 Intron XP_005266108.1
XM_011518157.2 31 Intron XP_011516459.1
XM_017014218.1 31 Intron XP_016869707.1
XM_017014219.1 31 Intron XP_016869708.1

View Full Product Details