Product Details

SNP ID
rs149279741
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96458437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGATGGAATGACAGCCGTGGGCC[A/G]GAGGGGATGCTCGAAAGAGCTGAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HABP4 PubMed Links

Gene Details

Gene
HABP4
Gene Name
hyaluronan binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014282.3 364 Silent Mutation CCA,CCG P136P NP_055097.2
XM_005251812.3 364 Silent Mutation CCA,CCG P26P XP_005251869.1
XM_017014484.1 364 UTR 5 XP_016869973.1

View Full Product Details