Product Details

SNP ID
rs149280437
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97428609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCAAACAGCTAGGTTTCCCTAC[A/G]CTAGAAGCCTATCTGAGAAGTGTGC
Phenotype
MIM: 611258
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TDRD7 PubMed Links

Gene Details

Gene
TDRD7
Gene Name
tudor domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302884.1 246 Intron NP_001289813.1
NM_014290.2 246 Silent Mutation ACA,ACG T48T NP_055105.2
XM_006717026.3 246 UTR 5 XP_006717089.1

View Full Product Details