Product Details

SNP ID
rs149678796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:83748340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGAAGCATTTTCAATAATTG[G/T]GCATTTCTTGCCTTTACTTCCTTAT
Phenotype
MIM: 611356
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GKAP1 PubMed Links

Gene Details

Gene
GKAP1
Gene Name
G kinase anchoring protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135953.1 596 Silent Mutation GCA,GCC A240A NP_001129425.1
NM_025211.3 596 Silent Mutation GCA,GCC A291A NP_079487.2
XM_005252241.2 596 Silent Mutation GCA,GCC A291A XP_005252298.1
XM_006717293.1 596 Silent Mutation GCA,GCC A167A XP_006717356.1
XM_011519058.2 596 Silent Mutation GCA,GCC A291A XP_011517360.1
XM_017015171.1 596 Silent Mutation GCA,GCC A167A XP_016870660.1

View Full Product Details