Product Details

SNP ID
rs149753101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:2804404 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGGTTTTTTCCAATGTAGGAATC[A/G]AGCTTTTCAGTGCAGCTTTGACTTT
Phenotype
MIM: 609960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PUM3 PubMed Links

Gene Details

Gene
PUM3
Gene Name
pumilio RNA binding family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014878.4 1970 Missense Mutation TCG,TTG S625L NP_055693.4

View Full Product Details