Product Details

SNP ID
rs149907184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96755659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGTATTATGGTTCCTATACAT[A/G]CCAGTCCTATTCACAAAATGATGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF510 PubMed Links

Gene Details

Gene
ZNF510
Gene Name
zinc finger protein 510
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314059.1 5443 UTR 3 NP_001300988.1
NM_001314060.1 5443 UTR 3 NP_001300989.1
NM_014930.2 5443 UTR 3 NP_055745.1
XM_005251808.2 5443 UTR 3 XP_005251865.1
XM_011518393.2 5443 UTR 3 XP_011516695.1
XM_017014483.1 5443 UTR 3 XP_016869972.1

View Full Product Details