Product Details

SNP ID
rs150099858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96458393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTTGGTAGGCCAGAAGCGGACT[C/T]CTAGAAGAGGGGAGCAGCAAGGATG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HABP4 PubMed Links

Gene Details

Gene
HABP4
Gene Name
hyaluronan binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014282.3 320 Missense Mutation CCT,TCT P122S NP_055097.2
XM_005251812.3 320 Missense Mutation CCT,TCT P12S XP_005251869.1
XM_017014484.1 320 UTR 5 XP_016869973.1

View Full Product Details