Product Details
- SNP ID
-
rs150328766
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:109379566 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTCACAGTTGTCTTAACTAGGA[C/T]CCAGCATTTGGACTAAACCTTCTTC
- Phenotype
-
MIM: 176877
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PTPN3
PubMed Links
Gene Details
- Gene
- PTPN3
- Gene Name
- protein tyrosine phosphatase, non-receptor type 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001145368.1 |
3584 |
Missense Mutation |
GAT,GGT |
D866G |
NP_001138840.1 |
NM_001145369.1 |
3584 |
Missense Mutation |
GAT,GGT |
D780G |
NP_001138841.1 |
NM_001145370.1 |
3584 |
Missense Mutation |
GAT,GGT |
D735G |
NP_001138842.1 |
NM_001145371.1 |
3584 |
Missense Mutation |
GAT,GGT |
D624G |
NP_001138843.1 |
NM_001145372.1 |
3584 |
Missense Mutation |
GAT,GGT |
D579G |
NP_001138844.1 |
NM_002829.3 |
3584 |
Missense Mutation |
GAT,GGT |
D911G |
NP_002820.3 |
XM_006717197.3 |
3584 |
Missense Mutation |
GAT,GGT |
D922G |
XP_006717260.1 |
XM_006717199.3 |
3584 |
Missense Mutation |
GAT,GGT |
D911G |
XP_006717262.1 |
XM_006717201.3 |
3584 |
Missense Mutation |
GAT,GGT |
D802G |
XP_006717264.1 |
XM_006717202.3 |
3584 |
Missense Mutation |
GAT,GGT |
D796G |
XP_006717265.1 |
XM_006717203.3 |
3584 |
Missense Mutation |
GAT,GGT |
D796G |
XP_006717266.1 |
XM_006717204.3 |
3584 |
Missense Mutation |
GAT,GGT |
D780G |
XP_006717267.1 |
XM_011518888.2 |
3584 |
Missense Mutation |
GAT,GGT |
D932G |
XP_011517190.1 |
XM_011518889.2 |
3584 |
Missense Mutation |
GAT,GGT |
D911G |
XP_011517191.1 |
XM_017014955.1 |
3584 |
Missense Mutation |
GAT,GGT |
D911G |
XP_016870444.1 |
XM_017014956.1 |
3584 |
Missense Mutation |
GAT,GGT |
D757G |
XP_016870445.1 |
XM_017014957.1 |
3584 |
Missense Mutation |
GAT,GGT |
D735G |
XP_016870446.1 |
View Full Product Details