Product Details

SNP ID
rs150472714
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:92300620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTCTCAAAGTTAGAAAAACTAT[C/T]GTTACTATTTACATCATGCCAACTC
Phenotype
MIM: 600709 MIM: 611534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IARS PubMed Links

Gene Details

Gene
IARS
Gene Name
isoleucyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
MIR3651
Gene Name
microRNA 3651
There are no transcripts associated with this gene.

Gene
NOL8
Gene Name
nucleolar protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256394.1 Intron NP_001243323.1
NM_017948.5 Intron NP_060418.4
XM_006717166.3 Intron XP_006717229.1
XM_006717167.3 Intron XP_006717230.1
XM_006717168.3 Intron XP_006717231.1
XM_006717169.3 Intron XP_006717232.1
XM_006717170.3 Intron XP_006717233.1
XM_006717172.3 Intron XP_006717235.1
XM_006717173.3 Intron XP_006717236.1
XM_011518824.2 Intron XP_011517126.1
XM_011518827.2 Intron XP_011517129.1
XM_017014876.1 Intron XP_016870365.1
XM_017014877.1 Intron XP_016870366.1
XM_017014878.1 Intron XP_016870367.1
XM_017014879.1 Intron XP_016870368.1
XM_017014880.1 Intron XP_016870369.1
Gene
SNORA84
Gene Name
small nucleolar RNA, H/ACA box 84
There are no transcripts associated with this gene.

View Full Product Details