Product Details

SNP ID
rs151084868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87967548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCCACGTGGAAGTCATGGATGT[C/G]GGGGGGCCCTGGATGGGCCTTGGGG
Phenotype
MIM: 610076
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDK20 PubMed Links

Gene Details

Gene
CDK20
Gene Name
cyclin dependent kinase 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039803.2 1265 Missense Mutation CAC,GAC H319D NP_001034892.1
NM_001170639.1 1265 UTR 3 NP_001164110.1
NM_001170640.1 1265 UTR 3 NP_001164111.1
NM_012119.4 1265 Missense Mutation CAC,GAC H298D NP_036251.2
NM_178432.3 1265 Missense Mutation CAC,GAC H311D NP_848519.1
XM_017014561.1 1265 Missense Mutation CAC,GAC H324D XP_016870050.1
XM_017014562.1 1265 UTR 3 XP_016870051.1
XM_017014563.1 1265 Missense Mutation CAC,GAC H283D XP_016870052.1
XM_017014564.1 1265 UTR 3 XP_016870053.1

View Full Product Details