Product Details

SNP ID
rs138038721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1341786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCGATGGTCCTCCTTTGGCTCAC[C/G]CTGCTCCTGATCGCCCTGCCCTGTC
Phenotype
MIM: 308385 MIM: 430000
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IL3RA PubMed Links

Gene Details

Gene
IL3RA
Gene Name
interleukin 3 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267713.1 408 Silent Mutation ACC,ACG T7T NP_001254642.1
NM_002183.3 408 Silent Mutation ACC,ACG T7T NP_002174.1
XM_005274431.4 408 Silent Mutation ACC,ACG T7T XP_005274488.1
XM_005274432.1 408 Silent Mutation ACC,ACG T7T XP_005274489.1
XM_017029491.1 408 Silent Mutation ACC,ACG T7T XP_016884980.1

View Full Product Details