Product Details

SNP ID
rs138173648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55488448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGTCTAGGTCACCAGAGCAGCT[C/T]CTCCGTGTCTGAGGCCTGGACCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP51 PubMed Links

Gene Details

Gene
USP51
Gene Name
ubiquitin specific peptidase 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201286.3 571 Silent Mutation AGA,AGG R164R NP_958443.1
XM_017029299.1 571 Silent Mutation AGA,AGG R164R XP_016884788.1
XM_017029300.1 571 Silent Mutation AGA,AGG R164R XP_016884789.1
XM_017029301.1 571 Silent Mutation AGA,AGG R164R XP_016884790.1

View Full Product Details