Product Details

SNP ID
rs138297441
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44844648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCGTGCGCATGATCAACTCGCC[G/T]GTAGGTAACATCCCTGACATCTATG
Phenotype
MIM: 300678
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DUSP21 PubMed Links

Gene Details

Gene
DUSP21
Gene Name
dual specificity phosphatase 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022076.3 646 Silent Mutation CCG,CCT P172P NP_071359.3

View Full Product Details