Product Details
- SNP ID
-
rs138602988
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:37694490 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCAGGCTTTCTTGGGCTCAGCCCC[A/C]CAGCTGACCCTACAGCTGTACATAA
- Phenotype
-
MIM: 314850
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
XK
PubMed Links
Gene Details
- Gene
- XK
- Gene Name
- X-linked Kx blood group
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
XM_011543978.2 |
646 |
Silent Mutation |
CCA,CCC |
P150P |
XP_011542280.1 |
View Full Product Details