Product Details

SNP ID
rs138804520
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46574585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCGTGCCCGGGCCGAGGTGGG[C/T]CTCGTCGAGGACACCGCCTGCGAGC
Phenotype
MIM: 300375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST7 PubMed Links

Gene Details

Gene
CHST7
Gene Name
carbohydrate sulfotransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019886.3 899 Silent Mutation GGC,GGT G218G NP_063939.2

View Full Product Details