Product Details

SNP ID
rs139334870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106941598 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACAGAAGTAAGGAGATAGCTGA[C/T]GTGGATACGTAGCCGCGTAAGCTTT
Phenotype
MIM: 300970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORC4 PubMed Links

Gene Details

Gene
MORC4
Gene Name
MORC family CW-type zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085354.2 2932 Missense Mutation CAT,CGT H886R NP_001078823.1
NM_024657.4 2932 Missense Mutation ATC,GTC I899V NP_078933.3
XM_005262190.3 2932 Missense Mutation ATC,GTC I807V XP_005262247.1
XM_006724691.2 2932 Missense Mutation ATC,GTC I777V XP_006724754.1
XM_011531027.2 2932 Missense Mutation ATC,GTC I722V XP_011529329.1
XM_017029844.1 2932 Missense Mutation CAT,CGT H794R XP_016885333.1

View Full Product Details