Product Details

SNP ID
rs140064980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119873342 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATTTTGGGTATCACTGGAGTCT[C/G]ATGGAAAGAGATAGGCGCATCTCTG
Phenotype
MIM: 300078 MIM: 300951
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA1 PubMed Links

Gene Details

Gene
NDUFA1
Gene Name
NADH:ubiquinone oxidoreductase subunit A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004541.3 282 Silent Mutation CTC,CTG L47L NP_004532.1
Gene
RNF113A
Gene Name
ring finger protein 113A
There are no transcripts associated with this gene.

View Full Product Details