Product Details

SNP ID
rs140187043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46853811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTTCCTCAAATATCAAATTTGG[A/G]TGTTTTCAATGGTACTATCCTGAAT
Phenotype
MIM: 300757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RP2 PubMed Links

Gene Details

Gene
RP2
Gene Name
retinitis pigmentosa 2 (X-linked recessive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006915.2 627 Silent Mutation GGA,GGG G146G NP_008846.2

View Full Product Details