Product Details

SNP ID
rs140764539
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30560111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATTGGGGTTTGTCTGCAGCACT[G/T]TGACTCTCTGACTTCTGCTATGTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CXorf21 PubMed Links

Gene Details

Gene
CXorf21
Gene Name
chromosome X open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025159.2 640 Missense Mutation AAA,ACA K82T NP_079435.1

View Full Product Details