Product Details

SNP ID
rs140765772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73213392 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGAAAAAATTGAAAAATGAATC[A/C]TTGGGAAAATCTTCAGTTACAGTTC
Phenotype
MIM: 300026
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAP1L2 PubMed Links

Gene Details

Gene
NAP1L2
Gene Name
nucleosome assembly protein 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021963.3 1483 Silent Mutation NP_068798.1

View Full Product Details