Product Details

SNP ID
rs141363402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:23000618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATAGAATCCAAAGCAACAAGCTG[C/T]ATGTCTGAAATGCAGGTGATTAACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX53 PubMed Links

Gene Details

Gene
DDX53
Gene Name
DEAD-box helicase 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182699.3 658 Silent Mutation TGC,TGT C187C NP_874358.2
Gene
PTCHD1-AS
Gene Name
PTCHD1 antisense RNA (head to head)
There are no transcripts associated with this gene.

View Full Product Details