Product Details

SNP ID
rs141412757
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:101553120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGGAGCTAAAACTAACGCTGGG[A/G]CAGGGTCTGGGGCCAAACTTCAGGG
Phenotype
MIM: 300362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARMCX1 PubMed Links

Gene Details

Gene
ARMCX1
Gene Name
armadillo repeat containing, X-linked 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016608.1 561 Missense Mutation ACA,GCA T64A NP_057692.1

View Full Product Details