Product Details

SNP ID
rs141414411
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:75379194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCAAGAGATGATGAGCCTTCTGG[A/G]TAATCTGCAAGGTTGAAACGTGAAG
Phenotype
MIM: 300576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZDHHC15 PubMed Links

Gene Details

Gene
ZDHHC15
Gene Name
zinc finger DHHC-type containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146256.1 1423 Silent Mutation TAC,TAT Y315Y NP_001139728.1
NM_001146257.1 1423 Intron NP_001139729.1
NM_144969.2 1423 Silent Mutation TAC,TAT Y324Y NP_659406.1
XM_006724624.3 1423 Silent Mutation TAC,TAT Y324Y XP_006724687.1
XM_017029296.1 1423 Intron XP_016884785.1
XM_017029297.1 1423 Silent Mutation TAC,TAT Y175Y XP_016884786.1
XM_017029298.1 1423 Intron XP_016884787.1

View Full Product Details