Product Details

SNP ID
rs141539936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53191935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGTAGGAGTTTGGGTGAGGGGG[C/T]CCGAGCCCATCTGTTTGGACTTGAG
Phenotype
MIM: 314690
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDM5C PubMed Links

Gene Details

Gene
KDM5C
Gene Name
lysine demethylase 5C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146702.1 6248 UTR 3 NP_001140174.1
NM_001282622.1 6248 UTR 3 NP_001269551.1
NM_004187.3 6248 UTR 3 NP_004178.2
XM_005262035.4 6248 Intron XP_005262092.1
XM_011530824.2 6248 Intron XP_011529126.1
XM_011530825.2 6248 Intron XP_011529127.1
XM_011530826.2 6248 Intron XP_011529128.1
XM_011530827.2 6248 Intron XP_011529129.1
XM_011530828.1 6248 Intron XP_011529130.1
XM_011530829.2 6248 Intron XP_011529131.1
XM_011530830.2 6248 Intron XP_011529132.1
XM_011530831.2 6248 Intron XP_011529133.1
XM_017029886.1 6248 Intron XP_016885375.1
XM_017029887.1 6248 Intron XP_016885376.1
XM_017029888.1 6248 Intron XP_016885377.1
XM_017029889.1 6248 Intron XP_016885378.1
Gene
MIR6894
Gene Name
microRNA 6894
There are no transcripts associated with this gene.

Gene
MIR6895
Gene Name
microRNA 6895
There are no transcripts associated with this gene.

View Full Product Details