Product Details

SNP ID
rs141687297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13777400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGGCAGAGCCACATATTTTTCC[A/G]GGGAAAGCATTCCAAGGAATGATAC
Phenotype
MIM: 300051 MIM: 300170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPM6B PubMed Links

Gene Details

Gene
GPM6B
Gene Name
glycoprotein M6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001994.2 1058 Silent Mutation NP_001001994.1
NM_001001995.2 1058 Silent Mutation NP_001001995.1
NM_001001996.2 1058 Silent Mutation NP_001001996.1
NM_001318729.1 1058 Silent Mutation NP_001305658.1
NM_005278.4 1058 Silent Mutation NP_005269.1
XM_005274489.4 1058 Silent Mutation XP_005274546.1
XM_011545497.2 1058 Silent Mutation XP_011543799.1
XM_017029432.1 1058 Silent Mutation XP_016884921.1
XM_017029433.1 1058 Silent Mutation XP_016884922.1
XM_017029434.1 1058 Silent Mutation XP_016884923.1
Gene
OFD1
Gene Name
OFD1, centriole and centriolar satellite protein
There are no transcripts associated with this gene.

View Full Product Details