Product Details

SNP ID
rs142497117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48698942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGTCTTCTTGGAATCAGCTGCA[A/G]GACCTGTGCCGCCTGGCCAAGCTCT
Phenotype
MIM: 300254 MIM: 300392
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUV39H1 PubMed Links

Gene Details

Gene
SUV39H1
Gene Name
suppressor of variegation 3-9 homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282166.1 359 Silent Mutation CAA,CAG Q31Q NP_001269095.1
NM_003173.3 359 Silent Mutation CAA,CAG Q20Q NP_003164.1
Gene
WAS
Gene Name
Wiskott-Aldrich syndrome
There are no transcripts associated with this gene.

View Full Product Details