Product Details

SNP ID
rs142591601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47585279 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACACCCCCGCCATGGAGAGTGT[C/G]TGCGGATACTTCCACAGGTCCCACA
Phenotype
MIM: 313440 MIM: 305370
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR4769 PubMed Links

Gene Details

Gene
MIR4769
Gene Name
microRNA 4769
There are no transcripts associated with this gene.

Gene
SYN1
Gene Name
synapsin I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006950.3 422 Intron NP_008881.2
NM_133499.2 422 Intron NP_598006.1
Gene
TIMP1
Gene Name
TIMP metallopeptidase inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003254.2 422 Silent Mutation GTC,GTG V92V NP_003245.1
XM_017029766.1 422 Missense Mutation TCT,TGT S16C XP_016885255.1

View Full Product Details